Skip to Content
Merck
All Photos(1)

Key Documents

TTR011

Sigma-Aldrich

Spirochetes TISSUE-TROL Control Slides

rabbit testicle tissue containing spirochetes (Treponema pallidum)

Sign Into View Organizational & Contract Pricing

Select a Size

25 EA
CZK 8,320.00

CZK 8,320.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 EA
CZK 8,320.00

About This Item

UNSPSC Code:
41116121
NACRES:
NA.47

CZK 8,320.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit testis (tissue containing spirochetes (Treponema pallidum))

application(s)

hematology
histology

storage temp.

room temp

Application

Histology slides with paraffin embedded section of rabbit testicle tissue containing spirochetes (Treponema pallidum). Suitable for use with Silver Stain, Modified Steiner-Steiner Procedure No. HT101, and other silver impregnation techniques.

Components

Box contains 24 unstained slides plus one reference slide stained using Modified Steiner-Steiner Silver Stain Procedure No. HT101.

Legal Information

TISSUE-TROL is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Is there anystained versions of the Spirochetes TISSUE-TROL™ Control Slides TTR011 for sale individually?

    1 answer
    1. The entire Tissue Trol product line was set up to provide 24 stained slides and 1 stained slide for each box of 25 slides. The intended market for these slides is clinical pathology labs. Tissue containing T pallidium is not easy to source. The only tissue where the tissue can be readily obtained are human and rabbit testes and armadillo foot pads. As such, obtaining tissue can be difficult.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service