Skip to Content
Merck

Skip To

T9656

Thymidine 5′-triphosphate sodium salt solution

≥99%, synthetic (organic), liquid

Synonym(s):

TTP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 μmol
Please contact Customer Service for Availability
CZK 3,210.00
0.1 mmol
Please contact Customer Service for Availability
CZK 10,100.00

About This Item

CAS Number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.52
MDL number:
Assay:
≥99%
Biological source:
synthetic (organic)
Form:
liquid
Storage temp.:
−20°C

CZK 3,210.00


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Thymidine 5′-triphosphate sodium salt solution, 100 mM in H2O

biological source

synthetic (organic)

Quality Segment

assay

≥99%

form

liquid

concentration

100 mM in H2O

color

colorless

foreign activity

DNase, RNase, none detected

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na].CC1=CN(C2CC(O)C(COP(O)(=O)OP(O)(=O)OP(O)(O)=O)O2)C(=O)NC1=O

InChI

1S/C10H17N2O14P3.Na.H/c1-5-3-12(10(15)11-9(5)14)8-2-6(13)7(24-8)4-23-28(19,20)26-29(21,22)25-27(16,17)18;;/h3,6-8,13H,2,4H2,1H3,(H,19,20)(H,21,22)(H,11,14,15)(H2,16,17,18);;

InChI key

KAELIXTVSANDSE-UHFFFAOYSA-N

General description

Thymidine 5′-triphosphate sodium salt solution is used in DNA synthesis reactions such as PCR, DNA sequencing, cDNA synthesis and molecular cloning techniques.

Application

Thymidine 5′-triphosphate sodium salt solution has been used as a component of the:
  • heat-sensitive solution for the preparation of a chemically defined diet that supports both larval development and adult survival of Drosophila[1]
  • dNTP mix for reverse transcription-polymerase chain reaction (RT-PCR) amplification
  • PCR for genomic DNA amplification by amplification refractory mutation system (ARMS) PCR

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
DTTP-ROT7791T0251
biological source

synthetic (organic)

biological source

-

biological source

-

biological source

-

Quality Level

200

Quality Level

-

Quality Level

200

Quality Level

200

assay

≥99%

assay

99% (dNTP)

assay

≥99%

assay

≥96%

form

liquid

form

solution

form

liquid

form

-

shipped in

dry ice

shipped in

-

shipped in

dry ice

shipped in

dry ice

concentration

100 mM in H2O

concentration

<0.9% (dNDP), 100 mM

concentration

10 mM (pH 7.0)

concentration

-


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters