T9571
Nunc® sealing tape for multiwell plates
polyolefin sealing tape, acrylate adhesive, non-pierceable
Synonym(s):
multiwell plate sealing films, sealing films
Select a Size
CZK 6,560.00
Select a Size
About This Item
CZK 6,560.00
Recommended Products
material
clear polyolefin film
description
not certified RNase/DNase-free
sterility
non-sterile
feature
piercable: no
packaging
pack of 100 ea
manufacturer/tradename
Nunc 232701
size
96 or 384 wells
color
clear
suitability
suitable for (microscopy, good optics, low auto-fluorescence, DMSO compatible)
Looking for similar products? Visit Product Comparison Guide
General description
Featuring 3M adhesive technology, sealing tapes provide a highly effective seal, preventing evaporation and eliminating contamination.
Specific tapes are intended for use in General Assays, Genomics, Compound and Library Storage, Bio-Analytical Assays, High Throughput Screening and Drug Discovery applications.
- Provide an effective seal on polypropylene, polystyrene, and polycarbonate microwell plates
- Compatible with aqueous solutions and organic solvents
- Tested for use between -80 and +110°C
- Pre-cut, plate-sized sheets
- Protective backing
- Minimal evaporation of plate contents
- Eliminates well-to-well cross contamination
- Easy to handle delivery system
- Provided with removable tabs
- For storage, PCR, microscopy, culture, protection
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service