T9014
β-Tetralone
Synonym(s):
3,4-Dihydro-2(1H)-naphthalenone, 3,4-Dihydro-2-naphthol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H10O
CAS Number:
Molecular Weight:
146.19
Beilstein:
509386
EC Number:
MDL number:
UNSPSC Code:
12352103
refractive index
n20/D 1.560 (lit.)
bp
131 °C/11 mmHg (lit.)
mp
18 °C (lit.)
density
1.106 g/mL at 25 °C (lit.)
SMILES string
O=C1CCc2ccccc2C1
InChI
1S/C10H10O/c11-10-6-5-8-3-1-2-4-9(8)7-10/h1-4H,5-7H2
InChI key
KCKZIWSINLBROE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
230.0 °F - closed cup
Flash Point(C)
110 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service