Skip to Content
Merck

T8950

Anti-TC10 antibody produced in rabbit

1 mg/mL, affinity isolated antibody, buffered aqueous solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
MDL number:
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
3
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human

concentration

1 mg/mL

technique(s)

western blot: 1 μg/mL using transfected COS cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RHOQ(23433)
mouse ... Rhoq(104215)

General description

TC10 has been classified as a member of the Rho family of GTPases and possesses characteristics most closely related to Cdc42. This family of proteins regulates a wide variety of cellular activity including cytoskeletal rearrangements, motility, prolifera

Immunogen

synthetic peptide corresponding to amino acid residues 136-152 from human TC10.

Biochem/physiol Actions

The antibody is specific for isotype TC10α and demonstrates slight cross-reactivity with TC10β.

Physical form

Solution in phosphate buffered saline containing 1.0 mg/mL BSA and 0.05 % sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library