Skip to Content
Merck
All Photos(1)

Key Documents

T8654

Magenta vessel

7-way tray

Synonym(s):

Magenta Vessel

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
CZK 2,730.00

CZK 2,730.00


Please contact Customer Service for Availability


Select a Size

Change View
10 EA
CZK 2,730.00

About This Item

UNSPSC Code:
41100000
NACRES:
NB.13

CZK 2,730.00


Please contact Customer Service for Availability

application(s)

agriculture

Looking for similar products? Visit Product Comparison Guide

General description

Culture tray supports eight GA-7 or GA-7-3 Magenta vessels or 36 25 mm culture tubes. Tray can be used during washing, filling, autoclaving or incubation

Legal Information

Magenta is a trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service