All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H7NO2S
CAS Number:
Molecular Weight:
157.19
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
Recommended Products
Product Name
L-α-(3-Thienyl)glycine,
Assay
≥98% (TLC)
form
powder
color
white
SMILES string
N[C@H](C(O)=O)c1ccsc1
InChI
1S/C6H7NO2S/c7-5(6(8)9)4-1-2-10-3-4/h1-3,5H,7H2,(H,8,9)/t5-/m0/s1
InChI key
BVGBBSAQOQTNGF-YFKPBYRVSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
X Liu et al.
Biopolymers, 33(8), 1237-1247 (1993-08-01)
Bradykinin (BK) is a peptide hormone with sequence Arg1-Pro2-Pro3-Gly4-Phe5-Ser6-Pro7-Phe8-Arg9 and has been implicated in a multitude of pathophysiological processes such as the ability to lower systemic blood pressure and stimulate pain. BK analogues having bulky, beta-branched D-aliphatic residues at position
J J Slon-Usakiewicz et al.
Biochemistry, 39(9), 2384-2391 (2000-03-01)
Synthetic bivalent thrombin inhibitors comprise an active site blocking segment, a fibrinogen recognition exosite blocking segment, and a linker connecting these segments. Possible nonpolar interactions of the P1' and P3' residues of the linker with thrombin S1' and S3' subsites
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service