Skip to Content
Merck
All Photos(1)

Key Documents

T8130

Sigma-Aldrich

9-(2′,3′,5′-Tri-O-benzyl-β-D-arabinofuranosyl)adenine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H31N5O4
CAS Number:
Molecular Weight:
537.61
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

form

solid

storage temp.

−20°C

SMILES string

Nc1ncnc2n(cnc12)C3OC(COCc4ccccc4)C(OCc5ccccc5)C3OCc6ccccc6

InChI

1S/C31H31N5O4/c32-29-26-30(34-20-33-29)36(21-35-26)31-28(39-18-24-14-8-3-9-15-24)27(38-17-23-12-6-2-7-13-23)25(40-31)19-37-16-22-10-4-1-5-11-22/h1-15,20-21,25,27-28,31H,16-19H2,(H2,32,33,34)

InChI key

WRFWNCVDEVBPJK-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service