Skip to Content
Merck
All Photos(1)

Documents

T7642

Sigma-Aldrich

L-(+)-Threose

≥60%, syrup

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O4
CAS Number:
Molecular Weight:
120.10
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

form

syrup

optical activity

[α]20/D 11.0 to 17.0, 48 hr, c = 2% (w/v) in water

concentration

≥60%

storage temp.

2-8°C

SMILES string

O[C@@H]1COC(O)[C@H]1O

InChI

1S/C4H8O4/c5-2-1-8-4(7)3(2)6/h2-7H,1H2/t2-,3+,4?/m1/s1

InChI key

FMAORJIQYMIRHF-BCDHYOAOSA-N

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Alda Lisa Concia et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 15(15), 3808-3816 (2009-02-18)
Novel aldol addition reactions of dihydroxyacetone (DHA) and hydroxyacetone (HA) to a variety of aldehydes catalyzed by D-fructose-6-phosphate aldolase (FSA) are presented. In a chemical-enzymatic cascade reaction approach, 1-deoxynojirimycin and 1-deoxymannojirimycin were synthesized starting from (R)- and (S)-3-(N-Cbz-amino)-2-hydroxypropanal, respectively. Furthermore
María Ruiz et al.
The Journal of organic chemistry, 73(6), 2240-2255 (2008-02-28)
A general strategy for the synthesis of 1-deoxy-azasugars from a chiral glycine equivalent and 4-carbon building blocks is described. Diastereoselective aldol additions of metalated bislactim ethers to matched and mismatched erythrose or threose acetonides and intramolecular N-alkylation (by reductive amination
Jussi Kinnunen et al.
Journal of biomedical optics, 17(9), 97003-97003 (2012-09-15)
Extensive collagen cross-linking affects the mechanical competence of articular cartilage: it can make the cartilage stiffer and more brittle. The concentrations of the best known cross-links, pyridinoline and pentosidine, can be accurately determined by destructive high-performance liquid chromatography (HPLC). We
Luis Miguel Azofra et al.
Carbohydrate research, 358, 96-105 (2012-07-31)
The potential energy surfaces for the different configurations of the D-erythrose and D-threose (open-chain, α- and β-furanoses) have been studied in order to find the most stable structures in the gas phase. For that purpose, a large number of initial
Laurence Burroughs et al.
Chemical communications (Cambridge, England), 46(26), 4776-4778 (2010-05-21)
Esters of proteinogenic amino acids efficiently catalyse the formation of erythrose and threose under aqueous conditions in the highest yields and enantioselectivities yet reported. Remarkably while esters of (L)-proline yield (L)-carbohydrates, esters of (L)-leucine and (L)-alanine generate (D)-carbohydrates, offering the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service