Skip to Content
Merck

T6384

Thyroid powder

Grade III

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Porcine thyroid

type

Grade III

form

powder

composition

Iodine, 0.8-0.9%

storage temp.

2-8°C

InChI

1S/C15H11I4NO4.Na/c16-8-4-7(5-9(17)13(8)21)24-14-10(18)1-6(2-11(14)19)3-12(20)15(22)23;/h1-2,4-5,12,21H,3,20H2,(H,22,23);/q;+1/p-1/t12-;/m0./s1

InChI key

YDTFRJLNMPSCFM-YDALLXLXSA-M

Application

To study the effects of thyroid hormone in animal models, thyroid powder can be administered by mixing with food for animal uptake. Treatment with thyroid powder was shown to improve cardiac function flowing induced myocardial infractions in rat models.

Analysis Note

Without diluent.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library