Skip to Content
Merck
All Photos(1)

Key Documents

T5299

Sigma-Aldrich

Trifucosyllacto-N-hexaose a

from human milk, ≥90% (HPLC)

Synonym(s):

TFLNH(a)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C58H98N2O43
CAS Number:
Molecular Weight:
1511.39
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

biological source

human milk

Assay

≥90% (HPLC)

form

solid

storage temp.

−20°C

SMILES string

CC1OC(OC2C(O)C(O)C(CO)OC2OC3C(NC(C)=O)C(OC(CO)C3OC4OC(C)C(O)C(O)C4O)OC5C(O)C(COC6OC(CO)C(OC7OC(CO)C(O)C(O)C7O)C(OC8OC(C)C(O)C(O)C8O)C6NC(C)=O)OC(OC(C(O)CO)C(O)C(O)C=O)C5O)C(O)C(O)C1O

InChI

1S/C58H98N2O43/c1-13-27(71)34(78)39(83)53(89-13)98-46-23(11-66)95-52(26(60-17(5)68)48(46)101-58-50(38(82)32(76)21(9-64)93-58)103-55-41(85)36(80)29(73)15(3)91-55)102-49-33(77)24(96-57(43(49)87)97-44(19(70)7-62)30(74)18(69)6-61)12-88-51-25(59-16(4)67)47(100-54-40(84)35(79)28(72)14(2)90-54)45(22(10-65)94-51)99-56-42(86)37(81)31(75)20(8-63)92-56/h6,13-15,18-58,62-66,69-87H,7-12H2,1-5H3,(H,59,67)(H,60,68)

InChI key

YOTBLRKYLWEPOA-UHFFFAOYSA-N

Other Notes

A natural nonasaccharide containing the Lewis-X substructure.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lars Bode et al.
Thrombosis and haemostasis, 92(6), 1402-1410 (2004-12-08)
Excessive leukocyte infiltration causes severe tissue damage in a variety of inflammatory diseases. The initial step in leukocyte extravasation is mediated by selectins and oligosaccharides on their glycoconjugate ligands. Human milk is a rich source of lactose-derived oligosaccharides that are
G Strecker et al.
Glycoconjugate journal, 6(2), 169-182 (1989-01-01)
The structure of a nonasaccharide and of two decasaccharides isolated from human milk has been investigated by using methylation, fast atom bombardment mass spectrometry and 1H-/13C-nuclear magnetic resonance spectroscopy. The structures of these oligosaccharides were: trifucosyllacto-N-hexaose; Fuc alpha 1-2Gal beta
Anja Pfenninger et al.
Journal of the American Society for Mass Spectrometry, 13(11), 1331-1340 (2002-11-22)
Underivatized neutral oligosaccharides from human milk were analyzed by nano-electrospray ionization (ESI) using a quadrupole ion trap mass spectrometer (QIT-MS) in the negative-ion mode. Under these conditions neutral oligosaccharides are observed as deprotonated molecules [M-H]- with high intensity. CID-experiments of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service