Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C10H14N2O5
CAS Number:
Molecular Weight:
242.23
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
assay
≥95% radiochemical purity basis (HPLC)
form
aqueous ethanol solution
extent of labeling
20-25 Ci/mmol
mass shift
M+6
storage temp.
2-8°C
SMILES string
[3H]C([3H])([3H])C1=CN([C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)NC1=O
Packaging
Combi-vial
Physical form
Aqueous solution containing 2% ethanol
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.