Skip to Content
Merck
All Photos(1)

Key Documents

T4521

Sigma-Aldrich

Triphenylmethane

~90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3CH
CAS Number:
Molecular Weight:
244.33
Beilstein:
1909753
EC Number:
MDL number:
UNSPSC Code:
12352100

Assay

~90%

bp

358-359 °C/754 mmHg (lit.)

mp

92-94 °C (lit.)

density

1.014 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

c1ccc(cc1)C(c2ccccc2)c3ccccc3

InChI

1S/C19H16/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15,19H

InChI key

AAAQKTZKLRYKHR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service