Skip to Content
Merck

Skip To

T3915

Transferrin-biotin labeled human

lyophilized powder containing sodium citrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Please contact Customer Service for Availability
CZK 6,970.00

About This Item

CAS Number:
UNSPSC Code:
12352202
eCl@ss:
34058011
EC Number:
234-318-8
NACRES:
NA.83
MDL number:
Form:
powder
Biological source:
human

CZK 6,970.00


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

form

powder

composition

Protein, ~90% biuret

extent of labeling

4-8 mol d-biotin per mol transferrin

technique(s)

cell culture | mammalian: suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... TF(7018)

Application

Biotin-labeled transferrin is commonly used to monitor transferrin uptake by cells in vitro through addition to the cell culture medium. [1]

Physical form

Lyophilized powder containing sodium citrate

Preparation Note

Prepared from human holo-transferrin, coupled to biotin by an amide bond through an aminocaproyl spacer.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T8158T2036T1147
biological source

human

biological source

-

biological source

-

biological source

human plasma

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

form

powder

form

powder

form

powder

form

powder

UniProt accession no.

P02787

UniProt accession no.

P02787

UniProt accession no.

P02787

UniProt accession no.

P02787

Gene Information

human ... TF(7018)

Gene Information

human ... TF(7018)

Gene Information

human ... TF(7018)

Gene Information

human ... TF(7018)

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Storage Class

11 - Combustible Solids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. Does the Biotin-Transferrin (#T3915-5mg) bind to Transferrin receptor 1?

    1 answer
    1. This product has not been evaluated for binding to Transferrin receptor 1.

      Helpful?

  2. Hello, does the Biotin-Transferrin (#T3915-5MG) have iron bound to it?

    1 answer
    1. The iron content of this item is not tested according to the CoA. This product is prepared from human holo-transferrin which has iron.

      Helpful?

Reviews

No rating value

Active Filters