Skip to Content
Merck

Skip To

T3879

L-Tyrosinamide

≥98% (TLC)

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C9H12N2O2
CAS Number:
Molecular Weight:
180.20
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26
MDL number:


Product Name

L-Tyrosinamide,

assay

≥98% (TLC)

form

powder

color

white to off-white

SMILES string

N[C@@H](Cc1ccc(O)cc1)C(N)=O

InChI

1S/C9H12N2O2/c10-8(9(11)13)5-6-1-3-7(12)4-2-6/h1-4,8,12H,5,10H2,(H2,11,13)/t8-/m0/s1

InChI key

PQFMNVGMJJMLAE-QMMMGPOBSA-N



Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters