Skip to Content
Merck

T3445

TFLZn potassium salt

≥90% (TLC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H15KN2O5S
CAS Number:
Molecular Weight:
410.49
UNSPSC Code:
12171500
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥90% (TLC)

color

tan

storage temp.

−20°C

SMILES string

[K+].COc1cc(NS(=O)(=O)c2ccc(cc2)C([O-])=O)c3nc(C)ccc3c1

InChI

1S/C18H16N2O5S.K/c1-11-3-4-13-9-14(25-2)10-16(17(13)19-11)20-26(23,24)15-7-5-12(6-8-15)18(21)22;/h3-10,20H,1-2H3,(H,21,22);/q;+1/p-1

InChI key

YNGNXRJCOYOGHQ-UHFFFAOYSA-M

Biochem/physiol Actions

May be used to measure intracellular Zn++ levels

Analysis Note

The TFLZn-Zn complex is fluorescent, with an excitation at 380 nm and emission at 510 nm.

Disclaimer

Protect from light


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library