Skip to Content
Merck
All Photos(1)

Key Documents

T2788

Sigma-Aldrich

Trizma® hydrochloride solution

pH 7.6, 1 M

Synonym(s):

Tris hydrochloride solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:

grade

for molecular biology

sterility

0.2 μm filtered

form

solution

concentration

1 M

impurities

DNase, RNase, NICKase and protease, none detected

pH

7.6

application(s)

diagnostic assay manufacturing

SMILES string

Cl.NC(CO)(CO)CO

InChI

1S/C4H11NO3.ClH/c5-4(1-6,2-7)3-8;/h6-8H,1-3,5H2;1H

InChI key

QKNYBSVHEMOAJP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

The pH values of all buffers are temperature and concentration dependent. For Tris buffers, pH increases about 0.03 unit per degree C decrease in temperature, and decreases 0.03-0.05 unit per ten-fold dilution.
For precise applications, use a carefully calibrated pH meter with a glass/calomel combination electrode.

Other Notes

Prepared with pH-adjusted Biotechnology Performance Certified Trizma Base in 18 megohm water and 0.2 μm filtered.

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mikael Marttinen et al.
Neurobiology of disease, 124, 454-468 (2018-12-18)
No single-omic approach completely elucidates the multitude of alterations taking place in Alzheimer's disease (AD). Here, we coupled transcriptomic and phosphoproteomic approaches to determine the temporal sequence of changes in mRNA, protein, and phosphopeptide expression levels from human temporal cortical
Anne-Laure Hemonnot-Girard et al.
Brain, behavior, and immunity, 91, 404-417 (2020-11-17)
Microglia, the resident immune cells of the brain, have recently emerged as key players in Alzheimer Disease (AD) pathogenesis, but their roles in AD remain largely elusive and require further investigation. Microglia functions are readily altered when isolated from their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service