Skip to Content
Merck
All Photos(1)

Key Documents

T2143

Sigma-Aldrich

trans-11-Tetradecenyl acetate

~97%

Synonym(s):

(E)-11-TDA, E-11-TDA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H30O2
CAS Number:
Molecular Weight:
254.41
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

~97%

form

solid

functional group

ester

shipped in

ambient

storage temp.

−20°C

SMILES string

CC\C=C\CCCCCCCCCCOC(C)=O

InChI

1S/C16H30O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-18-16(2)17/h4-5H,3,6-15H2,1-2H3/b5-4+

InChI key

YJINQJFQLQIYHX-SNAWJCMRSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Pheromone for Platynota stultana and Spodoptera litura.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yongping Huang et al.
Journal of chemical ecology, 28(3), 533-539 (2002-04-12)
Individual analysis of the female sex pheromone of the adzuki bean borer, Ostrinia scapulalis, has shown that the sex pheromone of this species comprised (E)-11-tetradecenyl acetate (E11-14:OAc) and (Z)-11-tetradecenyl acetate (Z11-14:OAc) at variable blend ratios. The pheromone blend could be
Ray L Hong et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(22), 7779-7784 (2008-05-30)
The geographical mosaic theory of coevolution predicts that different local species interactions will shape population traits, but little is known about the molecular factors involved in mediating the specificity of these interactions. Pristionchus nematodes associate with different scarab beetles around
Chang Yeol Yang et al.
Journal of chemical ecology, 35(3), 342-348 (2009-02-18)
We tested differences in female pheromone production and male response in three species of the genus Adoxophyes in Korea. Females of all three species produced mixtures of (Z)-9-tetradecenyl acetate (Z9-14:OAc) and (Z)-11-tetradecenyl acetate (Z11-14:OAc) as major components but in quite
A-E Félix et al.
Journal of chemical ecology, 35(5), 618-623 (2009-05-12)
The relationship between pheromone composition and mitochondrial haplotype clades was investigated by coupling DNA analyses with pheromone identification and male mate searching behavior among different geographic populations of Busseola fusca. The within-population variations in pheromone blend were as great as
S P Foster
Archives of insect biochemistry and physiology, 43(3), 125-136 (2000-02-24)
Pheromone titer in moths is a product of three processes occurring in or at the surface of the pheromone gland: biosynthesis, release, and intraglandular degradation, of pheromone. Changes in titers of sex pheromone, the fatty acyl pheromone analog (FAPA), and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service