SYNS386200
Synthware™ long syringe needle
O.D. × L 0.6 mm × 200 mm , Bevel Tip, needle, female luer lock
Synonym(s):
Synthware S386200, stainless steel syringe needle, bevel tip needle
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
feature
needle female luer lock
packaging
pack of 1 ea
manufacturer/tradename
Kemtech S386200
O.D. × L
0.6 mm × 200 mm , Bevel Tip
Looking for similar products? Visit Product Comparison Guide
General description
Long Syringe Needles, #304 SS, OD: 0.6Mm, Length: 200mm
Application
kemetch; synthware; needle; extended length
Features and Benefits
Organic Chemistry
Legal Information
Synthware is a trademark of Kemtech America Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service