Skip to Content
Merck
All Photos(1)

Key Documents

SRP8051

Sigma-Aldrich

CD152 (human): FC (human) (non-lytic)

recombinant, expressed in CHO cells, ≥98% (SDS-PAGE)

Synonym(s):

CTLA-4, Cytotoxic T-lymphocyte-associated antigen 4, Hepatitis A virus cellular receptor 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CZK 2,780.00

CZK 2,780.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
CZK 2,780.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

CZK 2,780.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human

recombinant

expressed in CHO cells

Assay

≥98% (SDS-PAGE)

form

lyophilized

mol wt

monomer 39 kDa by calculation

packaging

pkg of 100 μg

impurities

<0.06 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CTLA4(1493)

General description

CD152 (CTLA-4 (cytotoxic T-lymphocyte associated protein 4)) and CD28 are structurally homologous molecules that are members of the immunoglobulin (Ig) gene superfamily. Both CD152 and CD28 are composed of a single Ig V-like extracellular domain, a transmembrane domain and an intracellular domain.[1]
CD152 and CD28 are both expressed on the cell surface as homodimers or as monomers.[1] CD152 was originally identified as a gene that was specifically expressed by cytotoxic T lymphocytes.[2] However, CD152 transcripts have since been found in both Th1 and Th2, and CD4+ and CD8+ T cell clones. Whereas, CD28 expression is constitutive on the surfaces of 95% of CD4+ T cells and 50% of CD8+ T cells and is down regulated upon T cell activation, CD152 expression is upregulated rapidly following T cell activation and peaks approximately 24 hours following activation. Although both CD152 and CD28 can bind to the same ligands, CD152 binds to B71 and B72 with 20-100-fold higher affinity than CD28.

Biochem/physiol Actions

CD152 (CTLA-4) and CD28, together with their ligands B7-1 and B7-2 (cluster of differentiation 80 and 86 respectively), constitute one of the dominant costimulatory pathways that regulate T and B cell responses.[3][2] CTLA-4 (cytotoxic T-lymphocyte associated protein 4) is known to prevent immune responses, and its deficiency causes fatal lymphoproliferation in mouse models. Overexpression of the gene is observed in rheumatoid arthritis. Mutation in CTLA-4 is associated with insulin-dependent (type 1) diabetes mellitus, Grave′s disease and Hashimoto′s thyroiditis.[2]

Physical form

Lyophilized from 0.2 μm-filtered solution in PBS.

Reconstitution

Reconstitute at 100 μg/mL in sterile PBS.

Other Notes

The extracellular domain of human CD152 [CTLA-4] (aa 37-160) is fused to the N-terminus of the Fc region of a mutant human IgG1.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CTLA-4 and autoimmunity: new insights into the dual regulator of tolerance.
Romo-Tena J, et al.
Autoimmunity Reviews, 12(12), 1171-1176 (2013)
T-cell regulation by CD28 and CTLA-4.
Alegre M L, et al.
Nature Reviews: Immunology, 1(3), 220-220 (2001)
Enhanced expression of CTLA-4 (CD152) on CD4+ T cells in HIV infection.
Steiner K, et al.
Clinical and Experimental Immunology, 115(3), 451-451 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service