Skip to Content
Merck
All Photos(1)

Documents

SRP6369

Sigma-Aldrich

AKR1D1 human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

3o5bred, Aldo-keto reductase family 1, CBAS2, SRD5B1, member D1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

biological source

human

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

liquid

mol wt

39.5 kDa (346 aa, 1-326 aa + His Tag)

packaging

pkg of 100 μg

concentration

0.5 mg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... AKR1D1(6718)

General description

AKR1D1 (aldo-keto reductase 1D1) is the only steroid 5β-reductase in humans. This enzyme shows major expression in liver and resides in cytoplasm. It is a member of the unique subfamily, AKR1D, of the aldo-keto reductase (AKR) superfamily, that are NAD(P)(H)-dependent oxidoreductases. AKR1D1 contains the characteristic (α/β)8 triosephosphate isomerase barrel in its core region, and has a molecular weight of 37kDa. Its C-terminal contains the active site residing on the β strands, and the ligand binding site is formed by long flexible loops.
Recombinant human AKR1D1 protein, fused to His-tag at N-terminus, was expressed in E.coli and purified by using conventional chromatography techniques.

Biochem/physiol Actions

AKR1D1 (aldo-keto reductase 1D1) catalyzes the reduction of Δ4-3-ketosteroids to 5β-dihydrosteroids in a stereospecific manner, and is responsible for inducing A/B cis-configuration of bile acids. It is a critical enzyme for steroid hormone metabolism (except estrogens) and bile acid biosynthesis. Mutations in this gene are linked with bile acid deficiency resulting in usually fatal neonatal hepatitis and cholestasis. Variants in this gene influence the expression of p450 enzymes, such as CYP3A4, CYP2C8, CYP2C9, CYP2C19, and CYP2B6, where up-regulation of AKR1D1 results in overexpression of these enzymes. Hence, this enzyme might indirectly be responsible for variation in drug metabolism or efficacy and adverse drug events in different individuals.

Physical form

0.5 mg/mL in 20 mM Tris-HCl buffer (pH 8.0) containing 1 mM DTT, 100 mM NaCl and 20% glycerol.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In-Depth Dissection of the P133R Mutation in Steroid 5?-Reductase (AKR1D1): A Molecular Basis of Bile Acid Deficiency.
Chen M et al
Biochemistry, 54(41), 6343-6351 (2015)
The rate-determining steps of aldo-keto reductases (AKRs), a study on human steroid 5?-reductase (AKR1D1).
Chen M et al
Chemico-Biological Interactions, 234, 360-365 (2015)
Genetic variation in aldo-keto reductase 1D1 (AKR1D1) affects the expression and activity of multiple cytochrome P450s.
Chaudhry AS et al
Drug Metabolism and Disposition, 41(8), 1538-1547 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service