HLA class II histocompatibility antigen gamma chain isoform b, also known as CD74, is a transmembrane protein which, on antigen presenting cells, is the invariant chain that chaperones MHC class II dimers from the endoplasmic reticulum to the cell surface. CD74 is expressed by cells of both T lymphocyte and B lymphocyte lineages. It is upregulated in several cancers and is also expressed by nonimmune cells during inflammation. Recombinant human CD74 protein, fused to His-tag at N-terminus, was expressed in E.coli and purified by using conventional chromatography techniques.
Physical form
0.5 mg/mL in 20 mM Tris-HCl buffer (pH 8.0) containing 0.1 M NaCl, 1 mM DTT and 30% glycerol.
To be transmitted by its mosquito vector, dengue virus (DENV) must infect midgut epithelial cells, replicate and disseminate into the hemocoel, and finally infect the salivary glands, which is essential for transmission. The extrinsic incubation period (EIP) is very relevant
The Journal of investigative dermatology, 135(11), 2775-2784 (2015-06-04)
Melanoma is believed to be a highly immunogenic tumor and recent developments in immunotherapies are promising. IFN-γ produced by immune cells has a crucial role in tumor immune surveillance; however, it has also been reported to be pro-tumorigenic. In the
The Journal of cell biology, 143(7), 1971-1980 (1998-12-29)
During fertilization in Chlamydomonas, gametes of opposite mating types interact with each other through sex-specific adhesion molecules on their flagellar surfaces. Flagellar adhesion brings the cell bodies of the gametes into close contact and initiates a signal transduction pathway in
The Journal of experimental medicine, 197(11), 1467-1476 (2003-06-05)
Macrophage migration inhibitory factor (MIF) accounts for one of the first cytokine activities to have been described, and it has emerged recently to be an important regulator of innate and adaptive immunity. MIF is an upstream activator of monocytes/macrophages, and
Frontiers in microbiology, 9, 2536-2536 (2018-11-09)
Accumulation of compatible solutes is a common stress response of microorganisms challenged by high osmolarity; it can be achieved either through synthesis or import. These processes have been intensively studied in Bacillus subtilis, where systems for the production of the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.