Skip to Content
Merck
All Photos(2)

Key Documents

SRP5275

Sigma-Aldrich

SIRT3, active, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

SIR2L3

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
CZK 15,200.00

CZK 15,200.00


Please contact Customer Service for Availability


Select a Size

Change View
10 μG
CZK 15,200.00

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.32

CZK 15,200.00


Please contact Customer Service for Availability

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

123-196 RLU/min·ng

mol wt

~66 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SIRT3(23410)

General description

SIRT3 is a member of the class I of the Sirtuin family that is characterized by a Sirtuin core domain. Sirtuin are grouped into four classes and are homologs to the Sir2 protein. SIRT3 is known to regulate the epigenetic gene silencing and suppresses recombination of rDNA. SIRT3 may function as an intracellular regulatory protein with mono-ADP-ribosyltransferase activity that is essential for its silencing function. SIRT3 regulates and maintains basal ATP levels in the cell. SIRT3 also modulates mitochondrial intermediary metabolism and fatty acid use during fasting (3).

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katarína Smolková et al.
Scientific reports, 10(1), 8677-8677 (2020-05-28)
Wild type mitochondrial isocitrate dehydrogenase (IDH2) was previously reported to produce oncometabolite 2-hydroxyglutarate (2HG). Besides, mitochondrial deacetylase SIRT3 has been shown to regulate the oxidative function of IDH2. However, regulation of 2HG formation by SIRT3-mediated deacetylation was not investigated yet.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service