Skip to Content
Merck
All Photos(2)

Key Documents

SRP5239

Sigma-Aldrich

TAOK3 (1-463), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

DKFZp666H245, DPK, FLJ31808, JIK, MAP3K18

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
CZK 9,380.00

CZK 9,380.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
CZK 9,380.00

About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

CZK 9,380.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

221-299 nmol/min·mg

mol wt

~89 kDa

technique(s)

inhibition assay: suitable

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... TAOK3(51347)

General description

TAOK3 or TAO kinase 3 is a member of the serine/threonine-protein kinase family that inhibits the basal activity of Jun kinase and is negatively regulated by epidermal growth factor (EGF). Overexpression of TAOK3 may activate the ERK1/ERK2 and JNK/SAPK targets. TAOK3 is highly expressed in peripheral blood leukocytes (PBLs), thymus, spleen, kidney, skeletal muscle, heart and liver.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jason T Yustein et al.
Oncogene, 22(40), 6129-6141 (2003-09-19)
The Sterile-20 or Ste20 family of serine/threonine kinases is a group of signaling molecules whose physiological roles within mammalian cells are just starting to be elucidated. Here, in this report we present the characterization of three human Ste20-like kinases with
E Tassi et al.
The Journal of biological chemistry, 274(47), 33287-33295 (1999-11-24)
Mammalian members related to Saccharomyces cerevisiae serine/threonine kinase STE20 can be divided into two subfamilies based on their structure and function. The PAK subfamily is characterized by an N-terminal p21-binding domain (also known as CRIB domain), a C-terminal kinase domain

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service