Skip to Content
Merck
All Photos(1)

Key Documents

SRP5182

Sigma-Aldrich

FRS3, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

FRS2β, FRS2B, MGC17167, SNT2

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
CZK 8,620.00

CZK 8,620.00


Please contact Customer Service for Availability


Select a Size

Change View
50 μG
CZK 8,620.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

CZK 8,620.00


Please contact Customer Service for Availability

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~87 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... FRS3(10817)

General description

FRS3 or fibroblast growth factor receptor substrate 3 is a peripheral plasma membrane protein that is a substrate for the fibroblast growth factor receptor. During FGF or NGF stimulation, FRS3 becomes tyrosine phosphorylated and then serves as a platform for the recruitment of multiple signaling proteins for activation of the Ras-MAP kinase signaling cascade. FRS3 is mainly expressed in fibroblast and myoblast cell lines and undergo robust tyrosine phosphorylation in response to several mitogenic ligands. Through interaction with Rho family of small GTPases, FRS3 has been implicated in the reorganization of the actin cytoskeleton and subsequent morphological changes in various cells.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lin Huang et al.
Biochemical and biophysical research communications, 324(3), 1011-1017 (2004-10-16)
The control of cellular responses with fibroblast growth factors and neurotrophins is mediated through membrane-linked docking proteins, SNT (suc1-binding neurotrophic target)-1/FRS2alpha and SNT-2/FRS2beta. ERK1/2 are members of the mitogen-activated protein kinase family that regulate diverse cellular activities in response to
H Xu et al.
The Journal of biological chemistry, 273(29), 17987-17990 (1998-07-11)
Fibroblast growth factors (FGFs) stimulate tyrosine phosphorylation of a membrane-anchored adapter protein, FRS2/SNT-1, promoting its association with Shp-2 tyrosine phosphatase and upstream activators of Ras. Using the yeast two-hybrid protein-protein interaction assay, we show that FRS2/SNT-1 and a newly isolated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service