Skip to Content
Merck
All Photos(1)

Key Documents

SRP5142

Sigma-Aldrich

STAT5, His tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

MGF, STAT5A

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~92 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... STAT5A(6776)

General description

STAT5 (signal transducers and activators of transcription 5) is a member of the STAT family and this protein is phosphorylated by receptor associated protein kinases in response to cytokines and growth factors. The differentiation of T helper (Th) cells is regulated by members of the STAT family of signaling molecules. In BCR-ABL-positive cells, STAT5 is constitutively activated by tyrosine phosphorylation. STAT5 activation results in upregulation of Bcl-XL and increased resistance to induction of apoptosis. STAT5 is involved in expression and growth hormone-mediated sexually dimorphic regulation of cytochrome P450 3A10/lithocholic acid 6 beta-hydroxylase.

Physical form

Supplied in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Wang et al.
Molecular and cellular biology, 16(11), 6141-6148 (1996-11-01)
Stat5 was initially identified as a prolactin-induced member of the signal transducer and activator of transcription (Stat) family in sheep. However, Stat5 is also activated in the response to a variety of cytokines. In mice, and possibly in other species
A Subramanian et al.
Nucleic acids research, 26(9), 2173-2178 (1998-06-20)
The level of expression of a number of sexually differentiated liver proteins is primarily determined by plasma growth hormone (GH). Adult males have a pulsatile profile of GH release, while females have a relatively steady-state pattern of GH release. An
Alexa S Green et al.
Science advances, 1(8), e1500221-e1500221 (2015-11-26)
Fms-like tyrosine kinase 3 internal tandem duplication (FLT3-ITD) is frequently detected in acute myeloid leukemia (AML) patients and is associated with a dismal long-term prognosis. FLT3 tyrosine kinase inhibitors provide short-term disease control, but relapse invariably occurs within months. Pim

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service