Skip to Content
Merck
All Photos(2)

Key Documents

SRP5072

Sigma-Aldrich

PTPN11 (246-593), active, GST tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

BPTP3, CFC, MGC14433, NS1, PTP-1D, PTP2C, SH-PTP2, SH-PTP3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

507-687 nmol/min·mg

mol wt

~69 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PTPN11(5781)

General description

Mammalian PTPases can be subdivided into 1 of 2 broad categories: transmembrane receptor PTPases and intracellular PTPases. PTPN11 is one of the 2 closely related mammalian intracellular PTPases whose sequences encode 2 tandem SRC homology 2 (SH2) domains that are located at the amino-terminal side of a single PTPase catalytic domain. This PTP is widely expressed in most tissues and plays a regulatory role in various cell signaling events that are important for a diversity of cell functions, such as mitogenic activation, metabolic control, transcription regulation, and cell migration

Physical form

Supplied in 20mM MOPS, pH 7.5, 50mM NaCl, 10mM glutathione, 0.25mM DTT, 0.1mM PMSF, 30% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Other Notes

Amino acids : 246 - 593

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Ahmad et al.
Proceedings of the National Academy of Sciences of the United States of America, 90(6), 2197-2201 (1993-03-15)
A cDNA encoding a nontransmembrane protein-tyrosine phosphatase (PTP; EC 3.1.3.48), termed PTP2C, was isolated from a human umbilical cord cDNA library. The enzyme contains a single phosphatase domain and two adjacent copies of the src homology 2 (SH2) domain at
U Dechert et al.
Human genetics, 96(5), 609-615 (1995-11-01)
A 2.1-kb cDNA probe encoding the human SH2-domain containing protein-tyrosine phosphatase SH-PTP2 (PTPN11) was hybridized to human metaphase chromosomes in three independent experiments. In each instance, hybridization was maximal to chromosome 12q24.1-q24.3. The presence of SH-PTP2 cDNA crosshybridizing sequences located

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service