Skip to Content
Merck
All Photos(1)

Documents

SRP3127

Sigma-Aldrich

NT-1/BCSF-3 human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

Cardiotrophin-like cytokine, Novel Neurtrophin-1/B-Cell Stimulating Factor-3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

22.4 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CLCF1(23529)

General description

Novel neurotrophin-1/B-cell stimulating factor-3 (NNT-1/BSF-3), also known as cardiotrophin-like cytokine factor 1 (CLCF1), is encoded by the gene mapped to human chromosome 11q13.2. The encoded protein is a new member of the gp130 cytokine family. Recombinant human NNT-1/BCSF-3 is a 22.4 kDa protein containing 199 amino acid residues.

Biochem/physiol Actions

Novel neurotrophin-1/B-cell stimulating factor-3 (NNT-1/BSF-3) has neurotrophic and B cell stimulatory effects.In addition, it is also implicated in neuroimmunoendocrine modulation of corticotroph function. BSF-3 helps in the embryonic development and differentiation of motor, sensory and autonomic neurons. It also facilitates the bone formation during development and in the ongoing remodeling of endochondral bone. The encoded protein binds and activates glycoprotein 130 (gp130) and leukemia inhibitory factor receptor member b (LIFR-b). This ligand and receptor interaction further induces downstream signaling events and activates Janus kinase 1 (JAK1)-signal transducer and activator of transcription protein 3 (STAT3) signaling pathway. In vitro, it supports the survival of chick embryo motor and sympathetic neurons. In mice, NNT-1/BCSF-3 induces serum amyloid A, causes body weight loss and B cell hyperplasia associated with increased in serum IgG and IgM.

Sequence

MLNRTGDPGP GPSIQKTYDL TRYLEHQLRS LAGTYLNYLG PPFNEPDFNP PRLGAETLPR ATVDLEVWRS LNDKLRLTQN YEAYSHLLCY LRGLNRQAAT AELRRSLAHF CTSLQGLLGS IAGVMAALGY PLPQPLPGTE PTWTPGPAHS DFLQKMDDFW LLKELQTWLW RSAKDFNRLK KKMQPPAAAV TLHLGAHGF

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cold-induced sweating syndrome including Crisponi syndrome.
Hahn A F and Boman H
GeneReviews(?) (2011)
Novel neurotrophin-1/B cell-stimulating factor-3 (NNT-1/BSF-3) / cardiotrophin-like cytokine (CLC)?a novel gp130 cytokine with pleiotropic functions
George V, et al.
Cytokine & Growth Factor Reviews, 15(5), 325-336 (2004)
G Senaldi et al.
Proceedings of the National Academy of Sciences of the United States of America, 96(20), 11458-11463 (1999-09-29)
We have identified a cytokine of the IL-6 family and named it novel neurotrophin-1/B cell-stimulating factor-3 (NNT-1/BSF-3). NNT-1/BSF-3 cDNA was cloned from activated Jurkat human T cell lymphoma cells. Its sequence predicts a 225-aa protein with a 27-aa signal peptide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service