Skip to Content
Merck
All Photos(1)

Key Documents

SRP0467

Sigma-Aldrich

BRG1 (1480-1603) His tag human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

MRD16, SMARCA4, SNF2, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 4, SWI2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CZK 17,900.00

CZK 17,900.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
CZK 17,900.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

CZK 17,900.00


Please contact Customer Service for Availability

biological source

human

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

15.5 kDa

packaging

pkg of 100 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SMARCA4(6597)

General description

Human BRG1, also known as SMARCA4 (GenBank Accession No. NM_1128849) amino-acids 1480 – 1603 with N-terminal HIS-tag, MW = 15.5 kDa, expressed in an E. coli expression system.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Curtis, C.D., et al.
Molecular and Cellular Biology (2012)
He H., et al.
Cell Cycle, 11 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service