SRP0432
PDE4D2 active human
recombinant, expressed in baculovirus infected Sf9 cells, ≥80% (SDS-PAGE)
Synonym(s):
DPDE3, PDE43, STRK1, phosphodiesterase 4D, cAMP-specific
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
12352200
NACRES:
NA.32
biological source
human
recombinant
expressed in baculovirus infected Sf9 cells
Assay
≥80% (SDS-PAGE)
form
aqueous solution
specific activity
≥13600 pmol/min-μg
mol wt
84 kDa
packaging
pkg of 5 μg
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... PDE4D(5144)
General description
Human PDE4D2 (GenBank Accession No. NM_001197221) amino acids 2-507 (end) with N-terminal GST-tag, MW=84 kDa, expressed in a Baculovirus infected Sf9 cell expression system.
Application
Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.
Unit Definition
One unit is defined as the amount of enzyme that will convert 1 pmole of 3′, 5′-cAMP to 5′-AMP per min at 37°C in a reaction buffer.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
George S Baillie et al.
The Biochemical journal, 404(1), 71-80 (2007-02-10)
Beta2-ARs (beta2-adrenoceptors) become desensitized rapidly upon recruitment of cytosolic beta-arrestin. PDE4D5 (family 4 cAMP-specific phosphodiesterase, subfamily D, isoform 5) can be recruited in complex with beta-arrestin, whereupon it regulates PKA (cAMP-dependent protein kinase) phosphorylation of the beta2-AR. In the present
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service