Skip to Content
Merck
All Photos(1)

Key Documents

SML3821

Sigma-Aldrich

CAS-12290-201

≥98% (HPLC)

Synonym(s):

3-((1-(2-chloroacetyl)azetidin-3-yl)methyl)-7-fluoroquinazolin-4(3H)-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
CZK 4,690.00
25 MG
CZK 18,800.00

CZK 4,690.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
CZK 4,690.00
25 MG
CZK 18,800.00

About This Item

Empirical Formula (Hill Notation):
C14H13ClFN3O2
Molecular Weight:
309.72
UNSPSC Code:
12352200
NACRES:
NA.21

CZK 4,690.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

Biochem/physiol Actions

CAS-12290-201, a fluoroquinazolinone compound, is cell penetrant, selective and potent chemical probe for deubiquitinating enzymes (DUB) VCPIP1. CAS-12290-201 covalently labels the catalytic cysteine of VCPIP1.
Cell penetrant, selective and potent chemical probe for deubiquitinating enzymes (DUB) VCPIP1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Accelerating inhibitor discovery for deubiquitinating enzymes
Nature Communications, 14(1), 686-686 (2023)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service