Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C11H20N2O4
CAS Number:
Molecular Weight:
244.29
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
protect from light
Pricing and availability is not currently available.
Quality Segment
assay
≥98% (HPLC)
form
powder
storage condition
protect from light
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
-10 to -25°C
Biochem/physiol Actions
Serotonin transporter (SERT) and neuronal nitric oxide synthase (nNOS) interaction blocker with antidepressant efficacy in vivo.
ZZL-7 is a dipeptide that disrupts the interaction between serotonin transporter (SERT) and neuronal nitric oxide synthase (nNOS) by targeting the groove between the αB helix and βB folding of the nNOS PDZ domain. ZZL-7 decreases 293T cellular SERT-nNOS complex level (post 2h incubation at 1 μM) and demonstrates antidepressant efficacy in wt, but not nNos-/- mice in vivo (10, 20, and 40 mg/kg via intragastric administration or 10 mg/kg via i.p.) without undesirable side effects to the animals.
ZZL-7 is a dipeptide that disrupts the interaction between serotonin transporter (SERT) and neuronal nitric oxide synthase (nNOS) by targeting the groove between the αB helix and βB folding of the nNOS PDZ domain. ZZL-7 decreases 293T cellular SERT-nNOS complex level (post 2h incubation at 1 μM) and demonstrates antidepressant efficacy in wt, but not nNos-/- mice in vivo (10, 20, and 40 mg/kg via intragastric administration or 10 mg/kg via i.p.) without undesirable side effects to the animals.
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.