Skip to Content
Merck

SML3654

Lockdown

≥97% (HPLC)

Synonym(s):

2,2-Bis[4-(3,4-dicarboxyphenoxy)phenyl]propane, 4,4′-[(1-Methylethylidene)bis(4,1-phenyleneoxy)]bis[1,2-benzenedicarboxylic acid], BTB 13888

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C31H24O10
CAS Number:
Molecular Weight:
556.52
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥97% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC(C1=CC=C(OC2=CC(C(O)=O)=C(C(O)=O)C=C2)C=C1)(C)C(C=C3)=CC=C3OC4=CC(C(O)=O)=C(C(O)=O)C=C4

InChI

1S/C31H24O10/c1-31(2,17-3-7-19(8-4-17)40-21-11-13-23(27(32)33)25(15-21)29(36)37)18-5-9-20(10-6-18)41-22-12-14-24(28(34)35)26(16-22)30(38)39/h3-16H,1-2H3,(H,32,33)(H,34,35)(H,36,37)(H,38,39)

InChI key

KJLPSBMDOIVXSN-UHFFFAOYSA-N

Biochem/physiol Actions

Cell permeable, potent, and selective allosteric inhibitor of phosphatase PPM1F
Lockdown (Cpd12) is a cell permeable, potent, and selective allosteric inhibitor of phosphatase PPM1F.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library