Skip to Content
Merck

Skip To

SML3576

Mandipropamid

≥98% (HPLC)

Synonym(s):

(±)-Chlorophenyl)-N-[2-[3-methoxy-4-(prop-2-ynyloxy)phenyl]ethyl]-2-(prop-2-ynyloxy)acetamide, 4-Chloro-N-[2-[3-methoxy-4-(2-propyn-1-yloxy)phenyl]ethyl]-α-(2-propyn-1-yloxy)benzeneacetamide, Mandi

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C23H22ClNO4
CAS Number:
Molecular Weight:
411.88
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

Clc1ccc(cc1)C(OCC#C)C(=O)NCCc2cc(c(cc2)OCC#C)OC

InChI

1S/C23H22ClNO4/c1-4-14-28-20-11-6-17(16-21(20)27-3)12-13-25-23(26)22(29-15-5-2)18-7-9-19(24)10-8-18/h1-2,6-11,16,22H,12-15H2,3H3,(H,25,26)

InChI key

KWLVWJPJKJMCSH-UHFFFAOYSA-N

Biochem/physiol Actions

Chemical inducers of proximity; specifically induces complex formation between PYR1 and ABI (Mandi CIP system); fungicide
Mandipropamid (Mandi) is an agrochemical extensively used as fungicide. Mandipropamid specifically induces complex formation between a sixfold mutant of the plant hormone receptor pyrabactin resistance 1 (PYR1) and abscisic acid insensitive (ABI). Mandipropamid is a highly cell permeable, low toxic chemical inducers of proximity in vivo. It induces rapid colocalization of proteins contained fused receptor domain PYRMandi and receiver domain ABI.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML2719SML3155SML1445
description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

-10 to -25°C

storage temp.

−20°C

storage temp.

-10 to -25°C

storage temp.

−20°C

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear (Warmed)

solubility

DMSO: 5 mg/mL, clear (warmed)

color

white to beige

color

white to beige

color

white to beige

color

white to beige


Still not finding the right product?

Explore all of our products under Mandipropamid


pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters