Skip to Content
Merck
All Photos(1)

Key Documents

SML3532

Sigma-Aldrich

BAY-091

≥98% (HPLC)

Synonym(s):

(R)-2-(3-Cyano-2-(2′-fluoro-3′-methylbiphenyl-4-yl)-1,7-naphthyridin-4-ylamino)butanoic acid, BAY 091, BAY091

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H21FN4O2
Molecular Weight:
440.47
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

N#CC1=C(N=C2C=NC=CC2=C1N[C@H](CC)C(O)=O)C3=CC=C(C=C3)C4=C(C(C)=CC=C4)F

Biochem/physiol Actions

BAY-091 is a potent and highly selective PIP4K2A inhibitor (IC50 = 1.3/2.6 nM with 10/250 μM [ATP] by ADP-Glo, 8.5/16.4 nM with 10 μM/2 mM [ATP] by HTRF) with good cellular permeability (target engagement EC50 = 1.1 μM/intact cells vs 1.8 μM/cell lysate by CETSA) and no significant off-target activity as assessed by a 373-kinase panel (<60% inhibition at 1 μM).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lars Wortmann et al.
Journal of medicinal chemistry, 64(21), 15883-15911 (2021-10-27)
PIP4K2A is an insufficiently studied type II lipid kinase that catalyzes the conversion of phosphatidylinositol-5-phosphate (PI5P) into phosphatidylinositol 4,5-bisphosphate (PI4,5P2). The involvement of PIP4K2A/B in cancer has been suggested, particularly in the context of p53 mutant/null tumors. PIP4K2A/B depletion has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service