Skip to Content
Merck

Skip To

SML3449

Artepillin C analog A57

≥98% (HPLC), CREB-CRTC2 interaction inhibitor, powder

Synonym(s):

2-Oxazolidinone, 3-[(2E)-3-[1,1′-biphenyl]-3-yl-1-oxo-2-propen-1-yl]-4-phenyl-, (4S)- (ACI)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C24H19NO3
CAS Number:
Molecular Weight:
369.41
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Artepillin C analog A57, ≥98% (HPLC)

Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

Artepillin C analog 57 is a potent inhibitor of CREB-CRTC2 interaction that ameliorates metabolic syndrome in in obese mice. It suppresses CREB/CRTC2-mediated gluconeogenic and SREBP transcriptions.
Potent inhibitor of CREB-CRTC2 interaction that ameliorates metabolic syndrome in in obese mice


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters