Skip to Content

SML3443

PFI-653

≥98% (HPLC)

Synonym(s):

8-Oxa-2-azaspiro[4.5]dec-2-yl[2-[(2-pyrazinylmethyl)amino]-5-pyrimidinyl]methanone, Vanin-1-IN-1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H22N6O2
CAS Number:
Molecular Weight:
354.41
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

SMILES string

O=C(C1=CN=C(N=C1)NCC2=CN=CC=N2)N3CCC4(C3)CCOCC4

InChI

1S/C18H22N6O2/c25-16(24-6-1-18(13-24)2-7-26-8-3-18)14-9-21-17(22-10-14)23-12-15-11-19-4-5-20-15/h4-5,9-11H,1-3,6-8,12-13H2,(H,21,22,23)

InChI key

VGRLXWFIXGZRMS-UHFFFAOYSA-N

Biochem/physiol Actions

PFI-653 is a cell permeable, potent, and selective inhibitor of Vanin-1 (Vascular non-inflammatory molecule-1) enzyme.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library