Skip to Content
Merck

Skip To

SML3397

SRT3025 hydrochloride

≥98% (HPLC)

Synonym(s):

3025 HCl; 5-(3-Methoxypropyl)-2-phenyl-N-(2-(6-(pyrrolidin-1-ylmethyl)thiazolo[5,4-b]pyridin-2-yl)phenyl)thiazole-4-carboxamide, hydrochloride salt, 5-(3-Methoxypropyl)-2-phenyl-N-[2-[6-(1-pyrrolidinylmethyl)thiazolo[5,4-b]pyridin-2-yl]phenyl]-4-thiazolecarboxamide, hydrochloride salt, SRT 3025 HCl, SRT-3025 HCl, Sirtuin modulator 1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C31H31N5O2S2 · xHCl
Molecular Weight:
569.74 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

SMILES string

O=C(C1=C(CCCOC)SC(C2=CC=CC=C2)=N1)NC3=C(C4=NC5=CC(CN6CCCC6)=CN=C5S4)C=CC=C3.[xHCl]

Biochem/physiol Actions

Orally active sirtuin 1 (Sirt1) activator in cultures and in vivo.
SRT3025 is an orally active sirtuin 1 activator (Sirt1 EC1.5 ≤1 μM, Emax ≥150% and <300%) that reduces cellular acetylation of Sirt1 substrates FoxO1 (induced by RANKL ) and histone 3 (1-10 μM; murine bone marrow macrophages) and exhibits in vivo efficacy in a murine atherosclerosis model (3.18 g/kg high-cholesterol diet; mice) and ovariectomy-induced bone loss in female mice (50 & 100 mg/kg via daily p.o.).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML2784SML2880SML1138
description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

-10 to -25°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage condition

desiccated

storage condition

desiccated

storage condition

-

storage condition

desiccated

solubility

DMSO: 2 mg/mL, clear (Warmed)

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear (Warmed)

solubility

DMSO: 20 mg/mL, clear


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters