Skip to Content
Merck

Skip To

SML3391

Glatiramer acetate

≥95% (HPLC), Myelin basic protein mimetic, powder or Lyophilized powder or film

Synonym(s):

Cop 1 (polyamide), Cop-1, Copaxone, Copolymer 1, GA, Polymer with L-alanine, L-lysine, L-tyrosine, and L-glutamic acid acetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Please contact Customer Service for Availability
2 540,00 CZK
25 mg
Please contact Customer Service for Availability
10 300,00 CZK

About This Item

Linear Formula:
(C9H11NO3)a·(C6H14N2O2)b ·(C5H9NO4)c ·(C3H7NO2)d ·x(CH3CO2H)
UNSPSC Code:
12352200
NACRES:
NA.21
Assay:
≥95% (HPLC)
Form:
(Powder or Lyophilized powder or film)
Storage condition:
desiccated

2 540,00 CZK


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Glatiramer acetate, ≥95% (HPLC)

Quality Segment

assay

≥95% (HPLC)

form

(Powder or Lyophilized powder or film)

storage condition

desiccated

color

white to beige

storage temp.

-10 to -25°C

SMILES string

OC([C@@H](N)C)=O.OC(CC[C@H](N)C(O)=O)=O.NCCCC[C@H](N)C(O)=O.OC([C@@H](N)CC1=CC=C(C=C1)O)=O

InChI

1S/C9H11NO3.C6H14N2O2.C5H9NO4.C3H7NO2/c10-8(9(12)13)5-6-1-3-7(11)4-2-6;7-4-2-1-3-5(8)6(9)10;6-3(5(9)10)1-2-4(7)8;1-2(4)3(5)6/h1-4,8,11H,5,10H2,(H,12,13);5H,1-4,7-8H2,(H,9,10);3H,1-2,6H2,(H,7,8)(H,9,10);2H,4H2,1H3,(H,5,6)/t8-;5-;3-;2-/m0000/s1

InChI key

YLOCGHYTXIINAI-XKUOMLDTSA-N

Biochem/physiol Actions

Glatiramer acetate, a myelin basic protein mimetic, is an immunomodulator used for treatment of Multiple Sclerosis. Glatiramer acetate is a mixture of synthetic peptides randomly composed of glutamic acid, lysine, alanine, and tyrosine. It reduces relapse rate and disease progression in Multiple Sclerosis. Its mechanism of action is not fully understood. Glatiramer acetate generates anti-inflammatory Th2 cells, which produce neurotrophic factors.
Myelin basic protein mimetic; immunodmodulator; used to treat multiple sclerosis; generates anti-inflammatory Th2 cells, which produce neurotrophic factors; promotes neurogenesis

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML0748SML3497M1170
form

(Powder or Lyophilized powder or film)

form

powder

form

(Powder or Lyophilized powder or film)

form

powder

assay

≥95% (HPLC)

assay

≥95% (HPLC)

assay

≥95% (HPLC)

assay

≥90% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

200

storage temp.

-10 to -25°C

storage temp.

−20°C

storage temp.

-10 to -25°C

storage temp.

−20°C

storage condition

desiccated

storage condition

desiccated

storage condition

desiccated

storage condition

-

color

white to beige

color

white to beige

color

white to beige

color

white


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters