Skip to Content
Merck

Skip To

SML3380

Phenoxyethyl isobutyrate

≥95% (HPLC)

Synonym(s):

Propanoic acid, 2-methyl-, 2-phenoxyethyl ester (9CI, ACI)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C12H16O3
CAS Number:
Molecular Weight:
208.25
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥95% (HPLC)
Form:
oil
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

oil

color

, Colorless to light brown

storage temp.

2-8°C

SMILES string

O(CCOC(=O)C(C)C)c1ccccc1

InChI

1S/C12H16O3/c1-10(2)12(13)15-9-8-14-11-6-4-3-5-7-11/h3-7,10H,8-9H2,1-2H3

InChI key

MJTPMXWJHPOWGH-UHFFFAOYSA-N

Biochem/physiol Actions

Competitive antagonst of olfactory receptor OR2AT4
Phenoxyethyl isobutyrate is an antagonist of olfactory receptor OR2AT4, a G-protein-coupled receptor (GPCR) also expressed in human primary keratinocytes.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML3121SML3331SML1743
description

≥95% (HPLC)

description

≥95% (HPLC)

description

≥95% (HPLC)

description

≥95% (HPLC)

form

oil

form

oil

form

oil

form

oil

assay

≥95% (HPLC)

assay

≥95% (HPLC)

assay

≥95% (HPLC)

assay

≥95% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

color

, Colorless to light brown

color

colorless to yellow

color

colorless to brown

color

colorless to light yellow


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

278.9 °F - closed cup

flash_point_c

137.16 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters