Skip to Content

SML3347

NPSP795 hydrochloride

≥95% (HPLC)

Synonym(s):

(R)-3-(4-Cyano-3-(3-((1-(2,3-dihydro-1H-inden-2-yl)-2-methylpropan-2-yl)amino)-2-hydroxypropoxy)phenyl)propanoic acid hydrochloride, 4-Cyano-3-[(2R)-3-[[2-(2,3-dihydro-1H-inden-2-yl)-1,1-dimethylethyl]amino]-2-hydroxypropoxy]benzenepropanoic acid hydrochloride, SB-423562 hydrochloride, SHP635

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C26H32N2O4 · xHCl
Molecular Weight:
436.54 (free base basis)
UNSPSC Code:
41121801
NACRES:
NA.77
MDL number:
Assay:
≥95% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

SMILES string

O=C(O)CCC1=CC=C(C#N)C(OC[C@H](O)CNC(C)(C)CC2CC3=C(C=CC=C3)C2)=C1

InChI

1S/C26H32N2O4/c1-26(2,14-19-11-20-5-3-4-6-21(20)12-19)28-16-23(29)17-32-24-13-18(8-10-25(30)31)7-9-22(24)15-27/h3-7,9,13,19,23,28-29H,8,10-12,14,16-17H2,1-2H3,(H,30,31)/t23-/m1/s1

InChI key

NJBFJCJKWWIKRD-HSZRJFAPSA-N

Biochem/physiol Actions

Calcium-sensing receptor negative allosteric modulator; suppress airway hyperresponsiveness and inflammation in human cells and murine asthma surrogates
NPSP795 is a calcium-sensing receptor negative allosteric modulator (CaSR NAMs). NPSP795 suppress airway hyperresponsiveness and inflammation in human cells and murine asthma surrogates. It does not alter blood pressure, heart rate or serum calcium concentrations.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library