Skip to Content
Merck

Skip To

SML3191

Cotrimoxazole

≥98% (HPLC), Antibiotic, powder

Synonym(s):

4-Amino-N-(5-methyl-3-isoxazolyl)-benzenesulfonamide mixt. with 5-[(3,4,5-trimethoxyphenyl)methyl]-2,4-pyrimidinediamine, TMP/SMX, Trimethoprim/sulfamethoxazole

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Please contact Customer Service for Availability
2 160,00 CZK
25 mg
Please contact Customer Service for Availability
8 720,00 CZK

About This Item

Empirical Formula (Hill Notation):
C14H18N4O3·5C10H11N3O3S
CAS Number:
Molecular Weight:
1556.71
UNSPSC Code:
12352107
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder

2 160,00 CZK


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Cotrimoxazole, ≥98% (HPLC)

Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

[S](=O)(=O)(Nc4n[o]c(c4)C)c3ccc(cc3)N.Nc1nc(c(cn1)Cc2cc(c(c(c2)OC)OC)OC)N

InChI

1S/C14H18N4O3.C10H11N3O3S/c1-19-10-5-8(6-11(20-2)12(10)21-3)4-9-7-17-14(16)18-13(9)15;1-7-6-10(12-16-7)13-17(14,15)9-4-2-8(11)3-5-9/h5-7H,4H2,1-3H3,(H4,15,16,17,18);2-6H,11H2,1H3,(H,12,13)

InChI key

WZRJTRPJURQBRM-UHFFFAOYSA-N

Biochem/physiol Actions

Cotrimoxazole is a combination antibiotic consisting of one part trimethoprim to five parts sulfamethoxazole that is used to treat a variety of bacterial infections both Gram-positive and Gram-negative. Sulfamethoxazole inhibits bacterial dihydropteroate synthase while trimethoprim inhibits bacterial dihydrofolate reductase, thereby blocking the synthesis of nucleic acids.
combination antibiotic consisting of one part trimethoprim to five parts sulfamethoxazole

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A2487SML3579PZ0179
form

powder

form

liquid, ready-to-use solution

form

powder

form

powder

assay

≥98% (HPLC)

assay

-

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

room temp

solubility

DMSO: 2 mg/mL, clear

solubility

-

solubility

DMSO: 2 mg/mL, clear (Wramed)

solubility

DMSO: >25 mg/mL

color

white to beige

color

colorless to faint yellow

color

white to beige

color

white to off-white


Still not finding the right product?

Explore all of our products under Cotrimoxazole


pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. Bonjour. Dans la partie Description je peux lire "Cotrimoxazole is a combination antibiotic consisting of one part trimethoprim to five parts sulfamethoxazole". Vous pouvez me confirmer ce ratio 1:5 dans la ref SML3191 en particulier svp ? C'est pas 1:19?

    1 answer
    1. Yes, the material composition is a 1:5 ratio of trimethoprim to sulfamethoxazole. Please see the link below to a publication that may be helpful:
      https://link.springer.com/chapter/10.1007/978-3-642-81890-5_11#:~:text=Co%2Dtrimoxazole%20is%20a%20combination,in%20a%201%3A%205%20ratio.

      Helpful?

Reviews

No rating value

Active Filters