Skip to Content
Merck

Skip To

SML2960

Nizatidine

≥98% (HPLC)

Synonym(s):

Nizatidine, LY 139037, N′-[2-[[[2-[(Dimethylamino)methyl]-4-thiazolyl]methyl]thio]ethyl]-N-methyl-2-nitro-1,1-ethenediamine, N-{2-[2-(N,N-Dimethylaminomethyl)-4-thiazolylmethylthio]ethyl}-N′-methyl-2-nitro-1,1-ethenediamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 mg
Please contact Customer Service for Availability
962,00 CZK

About This Item

Empirical Formula (Hill Notation):
C12H21N5O2S2
CAS Number:
Molecular Weight:
331.46
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder

962,00 CZK


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[s]1c(nc(c1)CSCCN\C(=C\[N+](=O)[O-])\NC)CN(C)C

InChI

1S/C12H21N5O2S2/c1-13-11(6-17(18)19)14-4-5-20-8-10-9-21-12(15-10)7-16(2)3/h6,9,13-14H,4-5,7-8H2,1-3H3/b11-6+

InChI key

SGXXNSQHWDMGGP-IZZDOVSWSA-N

Biochem/physiol Actions

Nizatidine is a potent and selective antagonist of histamine H2 receptors used for treatment of stomach and intestine ulcers. Also, it was recommended as a component of combination therapy for cancer treatment. New data indicate that it can be repurposed to treat subtypes of clear cell renal cell carcinoma (ccRCC) in patients expressing certain isoforms of the pyruvate kinase muscle type. Additionally, nizatidine increases utrophin A levels in in C2C12 myoblasts (relevant to Duchenne muscular dystrophy).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
N1090000N70351467804
description

≥98% (HPLC)

description

European Pharmacopoeia (EP) Reference Standard

description

analytical standard

description

United States Pharmacopeia (USP) Reference Standard

form

powder

form

-

form

-

form

-

assay

≥98% (HPLC)

assay

-

assay

-

assay

-

Quality Level

100

Quality Level

-

Quality Level

100

Quality Level

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

storage temp.

-

solubility

DMSO: 2 mg/mL, clear

solubility

-

solubility

-

solubility

-

color

white to beige

color

-

color

-

color

-


Still not finding the right product?

Explore all of our products under Nizatidine


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

Flash Point (°F)

No data available

Flash Point (°C)

No data available



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters