Skip to Content
Merck

SML2458

M090

≥98% (HPLC)

Synonym(s):

(1R,2R,3R,5S)-N-((3-Cyclopropylthiophen-2-yl)methyl)-2,6,6-trimethylbicyclo[3.1.1]heptan-3-amine, 3-Cyclopropyl-N-[(1R,2R,3R,5S)-2,6,6-trimethylbicyclo[3.1.1]hept-3-yl]-2-thiophenemethanamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H27NS
CAS Number:
Molecular Weight:
289.48
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
oil
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

oil

color

colorless to very dark brown

storage temp.

2-8°C

SMILES string

C[C@H]1[C@H](NCC2=C(C3CC3)C=CS2)C[C@H]4C(C)(C)[C@@H]1C4

InChI

1S/C18H27NS/c1-11-15-8-13(18(15,2)3)9-16(11)19-10-17-14(6-7-20-17)12-4-5-12/h6-7,11-13,15-16,19H,4-5,8-10H2,1-3H3/t11-,13+,15-,16-/m1/s1

InChI key

MTOZOUMZDHSFBE-WXYCVUISSA-N

Biochem/physiol Actions

M090 is a potent inhibitor of influenza A hemagglutinin that is effective against amantadine-resistant and oseltamivir-resistant viruses.
TB Hemagglutinin (HA) inhibitor


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library