Skip to Content
Merck

Skip To

SML2398

SR16832

≥98% (HPLC)

Synonym(s):

2-Chloro-N-(6-methoxyquinolin-4-yl)-5-nitrobenzamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Please contact Customer Service for Availability
1 830,00 CZK

About This Item

Empirical Formula (Hill Notation):
C17H12ClN3O4
CAS Number:
Molecular Weight:
357.75
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder

1 830,00 CZK


Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

ClC1=CC=C([N+]([O-])=O)C=C1C(NC2=C(C=C(OC)C=C3)C3=NC=C2)=O

InChI

1S/C17H12ClN3O4/c1-25-11-3-5-15-13(9-11)16(6-7-19-15)20-17(22)12-8-10(21(23)24)2-4-14(12)18/h2-9H,1H3,(H,19,20,22)

InChI key

CVTZAGCRUDYUGB-UHFFFAOYSA-N

Biochem/physiol Actions

PPARγ antagonist
SR16832 is a PPARγ antagonist. It acts as a dual-site covalent inhibitor of PPARγ inhibiting both orthosteric and allosteric cellular activation by the PPARγ agonists.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters