Skip to Content
Merck

Skip To

SML2240

IU1-47

≥98% (HPLC)

Synonym(s):

1-[1-(4-Chlorophenyl)-2,5-dimethyl-1H-pyrrol-3-yl]-2-(1-piperidinyl)ethanone

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C19H23ClN2O
CAS Number:
Molecular Weight:
330.85
UNSPSC Code:
12352200
MDL number:
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder


assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

ClC1=CC=C(N2C(C)=CC(C(CN3CCCCC3)=O)=C2C)C=C1

InChI key

VXQIPRWKLACSKZ-UHFFFAOYSA-N

Biochem/physiol Actions

IU1-47 is a potent and selective inhibitor of the deubiquitinating enzyme UPS14. IU1-47 enhances protein degradation including human wild-type and pathological Tau forms in neurons.
potent and selective inhibitor of the deubiquitinating enzyme UPS14

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
I1911SML0662SML2226
assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: >10 mg/mL

solubility

DMSO: 5 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

color

white to beige

color

off-white to light brown

color

white to beige

color

white to beige


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters