Skip to Content
Merck

Skip To

SML2202

SGC-GAK-1

≥98% (HPLC)

Synonym(s):

6-Bromo-N-(3,4,5-trimethoxyphenyl)quinolin-4-amine, CA93

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Please contact Customer Service for Availability
2 630,00 CZK
2 235,50 CZK
25 mg
Please contact Customer Service for Availability
10 600,00 CZK
9 010,00 CZK

About This Item

Empirical Formula (Hill Notation):
C18H17BrN2O3
CAS Number:
Molecular Weight:
389.24
UNSPSC Code:
12352204
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder

2 235,50 CZK

List Price2 630,00 CZKSave 15%
Web-Only Promotion

Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

BrC1=CC2=C(NC3=CC(OC)=C(OC)C(OC)=C3)C=CN=C2C=C1

InChI

1S/C18H17BrN2O3/c1-22-16-9-12(10-17(23-2)18(16)24-3)21-15-6-7-20-14-5-4-11(19)8-13(14)15/h4-10H,1-3H3,(H,20,21)

InChI key

AUOSKLDNVNGKRR-UHFFFAOYSA-N

Biochem/physiol Actions

Non-cytotoxic, ATP-competitive cyclin G-associated kinase (GAK) inhibitor that effectively inhibits Dengue viral entry and packaging in cultures.
SGC-GAK-1 (CA93) is an ATP-competitive cyclin G-associated kinase (GAK) inhibitor (Ki = 3.1 nM by ATP site fluorescent tracer displacement assay; KD = 4.5 nM by ITC) that effectively inhibits Dengue viral entry and packaging in cultures (IC50 = 920 nM) without detectable cytotoxicity. SGC-GAK-1 exhibits much reduced affinity toward only 4 other kinases among a panel of ∼400 (KD = 1.9 nM/GAK vs. 110 nM/RIPK2, 190 nM/ADCK3, 520 nM/NLK, 980 nM/AVCR1 by ligand competitive binding assay; IC50 = 110 nM/GAK vs. 360 nM/RIPK2 by cellular target engagement assay). SGC-GAK-1N (CA71) is a structure analog and the recommended negative control (GAK KD = 7.1 μM by ligand competitive binding assay; GAK & RIPK2 IC50 >50 μM by cellular target engagement assay). For characterization details of SGC-GAK-1, please visit the SGC-GAK-1 probe summary on the Structural Genomics Consortium (SGC) website.

SGC-GAK-1N is the negative control for the active probe, SGC-GAK-1. SGC-GAK-1N is available from Sigma. To learn more about and purchase SGC-GAK-1N, click here.

To learn about other SGC chemical probes, visit sigma.com/sgc

Features and Benefits

SGC-GAK-1 is a chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC chemical probes, visit sigma.com/SGC.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML2203SML1358SML1960
description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 20 mg/mL, clear

solubility

DMSO: 20 mg/mL, clear

color

white to beige

color

dark green to black

color

white to light brown

color

white to beige


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters