Skip to Content
Merck

SML2000

3-BrOP

≥98% (GC)

Synonym(s):

3-BrPA propyl ester, 3-Bromo-2-oxopropionate-1-propyl ester, 3-Bromopyruvate propyl ester, Glycolycin, Propyl 3-bromo-2-oxopropanoate

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
C6H9BrO3
CAS Number:
Molecular Weight:
209.04
UNSPSC Code:
12352200
Assay:
≥98% (GC)
Form:
oil
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

assay

≥98% (GC)

form

oil

color

colorless to yellow

shipped in

ambient

storage temp.

2-8°C

Biochem/physiol Actions

3-BrOP is a cell-permeable ester of the the glycolysis inhibitor 3-bromopyruvate (3-BrPA) that is known to target hexokinase II (HK2) and GAPDH. Upon entering the cells, 3-BrOP is hydrolyzed by intracellular esterase to 3-BrPA. 3-BrOP is shown to synergize cancer cell killing with mTOR inhibitor rapamycin (apoptosis induction with/without 100 ng/mL rapamycin = 64%/36% of Raji human lymphoma cells by 40 μM 3-BrOP and 72%/45% of HL-60 leukemia cells by 20 μM 3-BrOP in 24 h) due to enhanced cellular ATP depletion and glucose uptake inhibition. When administered via tail vein injection, 3-BrOP significantly suppersses SK-N-SH xenograft-derived neuroblastoma tumor growth in mice in vivo (by 78% in 14 days; 20 mg/kg/day).
Cell-permeable ester of the the glycolysis inhibitor 3-bromopyruvate (3-BrPA) with in vitro and in vivo anti-cancer efficacy.

signalword

Warning

pictograms

Exclamation mark

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R-H Xu et al.
Leukemia, 19(12), 2153-2158 (2005-09-30)
The mammalian target of rapamycin (mTOR) pathway plays important roles in regulating nutrient metabolism and promoting the growth and survival of cancer cells, which exhibit increased glycolysis for ATP generation. In this study, we tested the hypothesis that inhibition of
Alejandro G Levy et al.
Investigational new drugs, 30(1), 191-199 (2010-10-05)
Children with high-risk and recurrent neuroblastoma have poor survival rates, and novel therapies are needed. Many cancer cells have been found to preferentially employ the glycolytic pathway for energy generation, even in the presence of oxygen. 3-BrOP is a novel
H-Q Ju et al.
Leukemia, 31(10), 2143-2150 (2017-02-15)
Internal tandem duplication (ITD) mutation in Fms-like tyrosine kinase 3 gene (FLT3/ITD) represents an unfavorable genetic change in acute myeloid leukemia (AML) and is associated with poor prognosis. Metabolic alterations have been involved in tumor progression and attracted interest as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service