Skip to Content
Merck

Skip To

SML1973

NVP-AEW541

≥98% (HPLC)

Synonym(s):

7-[cis-3-(1-Azetidinylmethyl)cyclobutyl]-5-[3-(phenylmethoxy)phenyl]-7H-pyrrolo[2,3-d]-pyrimidin-4-amine, AEW 541, AEW-541, AEW541, cis-7-(3-Aminomethyl-cyclobutyl)-5-(3-benzyloxy-phenyl)-7H-pyrrolo[2,3-d]-pyrimidin-4-ylamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Please contact Customer Service for Availability
4 460,00 CZK
3 791,00 CZK

About This Item

Empirical Formula (Hill Notation):
C27H29N5O
CAS Number:
Molecular Weight:
439.55
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder

3 791,00 CZK

List Price4 460,00 CZKSave 15%
Web-Only Promotion

Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

NC1=C2C(N([C@H]3C[C@@H](CN4CCC4)C3)C=C2C5=CC=CC(OCC6=CC=CC=C6)=C5)=NC=N1

InChI

1S/C27H29N5O/c28-26-25-24(21-8-4-9-23(14-21)33-17-19-6-2-1-3-7-19)16-32(27(25)30-18-29-26)22-12-20(13-22)15-31-10-5-11-31/h1-4,6-9,14,16,18,20,22H,5,10-13,15,17H2,(H2,28,29,30)

InChI key

AECDBHGVIIRMOI-UHFFFAOYSA-N

Biochem/physiol Actions

NVP-AEW541 is a reversible, ATP-competitive, orally available insulin-like growth factor-I receptor (IGF-IR, IGF-1R) inhibitor that, despite its equipotency against recombinant kinase domains of insulin receptor (InsR) and IGF-IR in cell-free kinase assays (IC50 = 140 & 150 nM, respectively), exhibits high selectivity toward cellular autophosphorylation of endogenous full-length IGF-1R (IC50 = 86 nM) over InsR (IC50 = 2.3 μM), HER1, PDGFR, c-Kit, and Bcr-Abl p210 (IC50 >5 μM). AEW541 effectively inhibits IGF-IR-mediated signaling and cancer survival (IC50 = 105 nM/MCF-7 soft agar growth, 1.64 μM/NWT-21 proliferation) in cultures and tumor growth in vivo (20 to 50 mg/kg twice daily p.o.; NWT-21 fibrosarcoma & TC-71 musculoskeletal sarcoma xenografts in mice).
Orally available insulin-like growth factor-I receptor inhibitor with in vitro and in vivo efficacy againsgt IGF-IR-mediated signaling and cancer survival.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML1921SML1107SML0314
assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 20 mg/mL, clear

solubility

DMSO: >10 mg/mL

color

white to beige

color

white to beige

color

white to beige

color

white to beige

7-[cis-3-(1-Azetidinylmethyl)cyclobutyl]-5-[3-(phenylmethoxy)phenyl]-7H-pyrrolo[2,3-d]-pyrimidin-4-amine

475488-23-4 NVP-ADW742, NVP-ADW742 10mg vial, NVP-ADW742 50mg for high-throughput screening, NVP-ADW742 FLNA silencing study, NVP-ADW742 IGF1R inhibitor, NVP-ADW742 Sigma-Aldrich, NVP-ADW742 cell proliferation assay reagent, NVP-ADW742 chemical structure, NVP-ADW742 cytotoxicity assay reagent, NVP-ADW742 for research, NVP-ADW742 insulin-like growth factor receptor inhibitor, NVP-ADW742 research chemical, high purity NVP-ADW742 for lab use

-

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters