Skip to Content
Merck

Skip To

SML1941

CGP3466B maleate salt

≥98% (HPLC)

Synonym(s):

CGP 3466B, Dibenz[b,f]oxepin-10-methanamine, N-methyl-N-2-propyn-1-yl-, (2Z)-2-butenedioate (1:1), N-(Dibenz[b,f]oxepin-10-ylmethyl)-N-methyl-N-(2-propynyl)amine monomaleic acid salt, Omigapil maleate salt, TCH346

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Please contact Customer Service for Availability
2 230,00 CZK
25 mg
Please contact Customer Service for Availability
9 000,00 CZK

About This Item

Empirical Formula (Hill Notation):
C19H17NO · C4H4O4
CAS Number:
Molecular Weight:
391.42
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder

2 230,00 CZK


Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

N(CC1=Cc2c(cccc2)Oc3c1cccc3)(CC#C)C.OC(=O)\C=C/C(=O)O

InChI

1S/C19H17NO.C4H4O4/c1-3-12-20(2)14-16-13-15-8-4-6-10-18(15)21-19-11-7-5-9-17(16)19;5-3(6)1-2-4(7)8/h1,4-11,13H,12,14H2,2H3;1-2H,(H,5,6)(H,7,8)/b;2-1-

InChI key

SQAZQLMBEHYFJA-BTJKTKAUSA-N

Biochem/physiol Actions

CGP3466B, also known as Omigapil maleate and TCH346, is a potent inhibitor of glyceraldehyde-3-phosphate dehydrogenase (GAPDH). It has been shown to have neuroprotective activity and received U.S. FDA Fast Track Designation for the treatment of congenital muscular dystrophy in May 2016.
Inhibitor of glyceraldehyde-3-phosphate dehydrogenase (GAPDH), neuroprotective.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
W108P5514Z0877
description

≥98% (HPLC)

description

powder

description

-

description

-

assay

≥98% (HPLC)

assay

-

assay

-

assay

>98% (HPLC)

form

powder

form

powder

form

powder

form

powder

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

-

storage temp.

-

storage temp.

room temp

solubility

DMSO: 2 mg/mL, clear

solubility

H2O: 25 mg/mL

solubility

-

solubility

DMSO: ≥20 mg/mL

color

white to beige

color

white

color

-

color

white to off-white


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters